Thermo Scientific Chemicals

Silicon lump, 10cm (3.9in) & down, 98.4% (metals basis), Thermo Scientific Chemicals

Catalog number: 010335.A7
5 kg, Each
Thermo Scientific Chemicals

Silicon lump, 10cm (3.9in) & down, 98.4% (metals basis), Thermo Scientific Chemicals

Catalog number: 010335.A7
5 kg, Each
Quantity
Catalog number: 010335.A7
also known as 010335-A7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-21-3
IUPAC Name
silicon
Molecular Formula
Si
InChI Key
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES
[Si]
Appearance (Color)
Silver to grey
Form
Lump or chunks
Assay (unspecified)
≥98.4%
Trace Metal
Aluminium (Al): ≤0.25%
Trace Metal
Calcium (Ca): ≤0.25%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text