Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 010336.06
also known as 010336-06
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
Add to cart
Sodium, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010336.06
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace Metal0.05%
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only