Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals
Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
4 x 250 g
Catalog number 010340.30
also known as 010340-30
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
Add to cart
Sodium, dispersion, 40% in oil, Thermo Scientific Chemicals
Catalog number010340.30
Price (USD)/ Each
108.65
Online exclusive
121.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
FormDispersion in oil
Appearance (Color)Grey
Assay from Supplier's CofASodium: 39.9-40.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only