Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010350.G1
also known as 010350-G1
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
-
Add to cart
Quantity:
1 m
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
Add to cart
Tantalum wire, 1.0mm (0.04in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010350.G1
Price (USD)/ Each
90.65
Online exclusive
100.00 
Save 9.35 (9%)
-
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only