Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals

Form: approx. 6.3g/100x150mm; Temper: as rolled
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
100 x 150 mm
150 x 500 mm
150 x 2000 mm
Catalog number 010351.FY
also known as 010351-FY
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
-
Add to cart
Quantity:
50 x 100 mm
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
Add to cart
Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010351.FY
Price (USD)/ Each
52.65
Online exclusive
57.90 
Save 5.25 (9%)
-
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormFoil, 0.025mm (0.001in) thick, hardened.
FormFoil, 0.025mm (0.001in) thick, hard.
Assay from Supplier's CofA99.95% (metals basis)
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only