Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Form: approx. 11.3g/100x100mm
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
100 x 500 mm
100 x 2000 mm
Catalog number 010385.GH
also known as 010385-GH
Price (USD)/ Each
67.65
Online exclusive
75.30 
Save 7.65 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
67.65
Online exclusive
75.30 
Save 7.65 (10%)
Add to cart
Titanium foil, 0.25mm (0.01in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number010385.GH
Price (USD)/ Each
67.65
Online exclusive
75.30 
Save 7.65 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Comment0.25mm thick
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only