Thermo Scientific Chemicals

Titanium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010386.36
Each
Thermo Scientific Chemicals

Titanium powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010386.36
Each
Catalog number: 010386.36
also known as 010386-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7440-32-6
IUPAC Name
titanium
Molecular Formula
Ti
InChI Key
RTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES
[Ti]
Particle Size
-325 mesh (less than 45 microns)
Appearance (Color)
Dark Grey
Trace Metals
less than 1% trace metals

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text