Titanium wire, 0.5mm (0.02in) dia, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Titanium wire, 0.5mm (0.02in) dia, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium wire, 0.5mm (0.02in) dia, annealed, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 cm
50 cm
250 cm
Catalog number 010389.CD
also known as 010389-CD
Price (USD)/ Each
834.00
-
Add to cart
Quantity:
1000 cm
Request bulk or custom format
Price (USD)/ Each
834.00
Add to cart
Titanium wire, 0.5mm (0.02in) dia, annealed, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number010389.CD
Price (USD)/ Each
834.00
-
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.99% (metals basis)
FormWire, 0.5mm (0.02in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only