Thermo Scientific Chemicals

Tungsten foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 010416.HW
150 x 300 mm, Each
Thermo Scientific Chemicals

Tungsten foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 010416.HW
150 x 300 mm, Each
Quantity
Catalog number: 010416.HW
also known as 010416-HW
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-33-7
IUPAC Name
tungsten
Molecular Formula
W
InChI Key
WFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES
[W]
Assay from Supplier's CofA
≥99.95% (metals basis)
Assay from Supplier's CofA
Ca: ≤ 0.003 %
Assay from Supplier's CofA
Cu: ≤ 0.002 %
Assay from Supplier's CofA
Mg: ≤ 0.002 %
Assay from Supplier's CofA
Ni: ≤ 0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text