Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
100 x 500 mm
Catalog number 010417.GH
also known as 010417-GH
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number010417.GH
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Assay from Supplier's CofAAl: ≤ 0.002 %
Assay from Supplier's CofACa: ≤ 0.003 %
Assay from Supplier's CofACu: ≤ 0.002 %
Assay from Supplier's CofAFe: ≤ 0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only