Cobalt powder, 1.6 micron, 98.8% (metals basis), Thermo Scientific Chemicals
Cobalt powder, 1.6 micron, 98.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt powder, 1.6 micron, 98.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Catalog number 010455.36
also known as 010455-36
Price (USD)/ Each
336.65
Online exclusive
374.00 
Save 37.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
336.65
Online exclusive
374.00 
Save 37.35 (10%)
Add to cart
Cobalt powder, 1.6 micron, 98.8% (metals basis), Thermo Scientific Chemicals
Catalog number010455.36
Price (USD)/ Each
336.65
Online exclusive
374.00 
Save 37.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only