Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, alpha-phase, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010459.22
100 g, Each
Thermo Scientific Chemicals

Aluminum oxide, gamma-phase, alpha-phase, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 010459.22
100 g, Each
Quantity
Catalog number: 010459.22
also known as 010459-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Total Metal Impurities
0.02% max.

Description

Aluminum oxide finds use as catalyst in organic synthesis and chromatography. It is used as an ingredient in sunscreen lotions and paints. It is also used in the manufacture of abrasives, refractories, ceramics, filler for plastics, dosimeter, spark plug insulators, and electrical insulators. Aluminum oxide is used widely to produce aluminum metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text