Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Thermo Scientific Chemicals

Catalog number: 010466.MD
250 mg, Each
Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Thermo Scientific Chemicals

Catalog number: 010466.MD
250 mg, Each
Quantity
Catalog number: 010466.MD
also known as 010466-MD
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12092-47-6
IUPAC Name
bis(λ¹-rhodium(1+)) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular Formula
C16H24Cl2Rh2
InChI Key
QSUDXYGZLAJAQU-MIXQCLKLSA-L
SMILES
[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Assay (unspecified)
Rh 40.5-42.1%

Description

It is a widely used precursor to homogeneous catalysts. This is a chiral catalyst capable of asymmetrically hydrogenating certain prochiral alkenes. Chloro(1,5-cyclooctadiene)rhodium(I) dimer is also used in the synthesis of other metal ligands for use in catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text