Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 010468.06
5 g, Each
Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals

Catalog number: 010468.06
5 g, Each
Quantity
Catalog number: 010468.06
also known as 010468-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14694-95-2
IUPAC Name
λ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular Formula
C54H45ClP3Rh
InChI Key
IXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES
[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
96.1% min

Description

Chlorotris(triphenylphosphine)rhodium(I) is used as a homogeneous hydrogenation catalyst. It is used in the selective hydrogenation of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text