Thermo Scientific Chemicals

trans-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals

Catalog number: 010472.03
1 g, Each
Thermo Scientific Chemicals

trans-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific Chemicals

Catalog number: 010472.03
1 g, Each
Quantity
Catalog number: 010472.03
also known as 010472-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14913-33-8
IUPAC Name
dichloroplatinum diamine
Molecular Formula
Cl2H6N2Pt
InChI Key
LXZZYRPGZAFOLE-UHFFFAOYSA-L
SMILES
N.N.Cl[Pt]Cl
Assay (unspecified)
Pt 64.5% min

Description

trans-Diamminedichloroplatinum(II) acts as a reversible RNA-protein cross-linking agent. It is also used in plating baths. It reacts with DNA after hydrolysis to get both intra and interstrand crosslinks. These crosslinks appear to impair replication and transcription of DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text