Thermo Scientific Chemicals

trans-Dichlorobis(diethylsulfide)platinum(II), Pt 43.7%, Thermo Scientific Chemicals

Catalog number: 010484.06
5 g, Each
Thermo Scientific Chemicals

trans-Dichlorobis(diethylsulfide)platinum(II), Pt 43.7%, Thermo Scientific Chemicals

Catalog number: 010484.06
5 g, Each
Quantity
Catalog number: 010484.06
also known as 010484-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15337-84-5
IUPAC Name
bis((ethylsulfanyl)ethane); dichloroplatinumbis(ylium)
Molecular Formula
C8H20Cl2PtS2
InChI Key
RFSLRYGKHBDFGC-UHFFFAOYSA-L
SMILES
Cl[Pt++]Cl.CCSCC.CCSCC
Assay (unspecified)
Pt 43.7% min

Description

It is used in Hydrosilylation. Platinum(II) complexes of diethylsulfide are convenient and versatile precursor for the preparation of a variety of organoplatinum(II) derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text