Thermo Scientific Chemicals

cis-Dichlorobis(triphenylphosphine)platinum(II), Pt 24.2% min, Thermo Scientific Chemicals

Catalog number: 010492.MD
250 mg, Each
Thermo Scientific Chemicals

cis-Dichlorobis(triphenylphosphine)platinum(II), Pt 24.2% min, Thermo Scientific Chemicals

Catalog number: 010492.MD
250 mg, Each
Quantity
Catalog number: 010492.MD
also known as 010492-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15604-36-1
IUPAC Name
dichloroplatinumbis(ylium); bis(triphenylphosphane)
Molecular Formula
C36H30Cl2P2Pt
InChI Key
ILIBUWCDNDIUOT-UHFFFAOYSA-L
SMILES
Cl[Pt++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
Pt 24.2% min

Description

cis-Dichlorobis(triphenylphosphine)platinum(II) is used primarily as a reagent for the synthesis of other platinum molecules and in platinum plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text