Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Thermo Scientific Chemicals
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 010494.03
also known as 010494-03
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, Thermo Scientific Chemicals
Catalog number010494.03
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS50982-12-2
IUPAC Nameλ²-ruthenium(2+) (1Z,5Z)-cycloocta-1,5-diene dichloride
Molecular FormulaC8H12Cl2Ru
InChI KeyDMRVBCXRFYZCPR-PHFPKPIQSA-L
SMILES[Cl-].[Cl-].[Ru++].C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ru 32% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only