Thermo Scientific™

Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010509.22
100 g, Each
Thermo Scientific™

Germanium(IV) chloride, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010509.22
100 g, Each
Quantity
Catalog number: 010509.22
also known as 010509-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10038-98-9
IUPAC Name
germanium tetrachloride
Molecular Formula
Cl4GeH8
InChI Key
VDHBHZZBEGQJRK-UHFFFAOYSA-N
SMILES
Cl.Cl.Cl.Cl.[GeH4]
Total Metal Impurities
0.0001% max.

Description

It is used as a catalyst in the conversion of carbohydrates into 5-hydroxymethylfurfural in ionic liquids, as a reducing agent in combination with triphenylphosphine (TPP) for the reduction of alpha-bromo carboxylic acid derivatives, and in the production of pure germanium. It also finds use as an intermediate for several optical processes. It is one of the most important dopants in silica glass for optical fibers. It is employed in the preparation of germanium dioxide which is used for wide camera lens, microscopy, IR-transparent glasses/windows/lenses, and for the core of fiber-optic lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text