Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals
Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 010510.06
also known as 010510-06
Price (USD)/ Each
77.65
Online exclusive
86.50 
Save 8.85 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
77.65
Online exclusive
86.50 
Save 8.85 (10%)
Add to cart
Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number010510.06
Price (USD)/ Each
77.65
Online exclusive
86.50 
Save 8.85 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-53-8
IUPAC Namegermanedione
Molecular FormulaGeO2
InChI KeyYBMRDBCBODYGJE-UHFFFAOYSA-N
SMILESO=[Ge]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
Germanium dioxide is used in algaculture.A mixture is used as an optical material for optical fibers and optical waveguides It is also used as a catalyst in production of polyethylene terephthalate resin and for production of other germanium compounds. It is used as a feedstock for production of some phosphors and semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only