Thermo Scientific Chemicals

Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010510.22
100 g, Each
Thermo Scientific Chemicals

Germanium(IV) oxide, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010510.22
100 g, Each
Quantity
Catalog number: 010510.22
also known as 010510-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-53-8
IUPAC Name
germanedione
Molecular Formula
GeO2
InChI Key
YBMRDBCBODYGJE-UHFFFAOYSA-N
SMILES
O=[Ge]=O
Total Metal Impurities
0.0001% max.

Description

Germanium dioxide is used in algaculture.A mixture is used as an optical material for optical fibers and optical waveguides It is also used as a catalyst in production of polyethylene terephthalate resin and for production of other germanium compounds. It is used as a feedstock for production of some phosphors and semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text