Thermo Scientific Chemicals

Hexaammineruthenium(III) chloride, Ru 32.1% min, Thermo Scientific Chemicals

Catalog number: 010511.01
0.25 g, Each
Thermo Scientific Chemicals

Hexaammineruthenium(III) chloride, Ru 32.1% min, Thermo Scientific Chemicals

Catalog number: 010511.01
0.25 g, Each
Quantity
Catalog number: 010511.01
also known as 010511-01
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14282-91-8
IUPAC Name
ruthenium(3+) hexaamine trichloride
Molecular Formula
Cl3H18N6Ru
InChI Key
GBDZMMXUOBAJMN-UHFFFAOYSA-K
SMILES
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3]
Assay (unspecified)
Ru 32.1% min

Description

Depending on the choice of suitable reductive or oxidative quencher this catalyst can be used to trigger photoreduction or photooxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text