Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals
Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
0.25 g
1 g
Catalog number 010514.06
also known as 010514-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS15635-87-7
IUPAC Nameiridium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21IrO6
InChI KeyHLYTZTFNIRBLNA-LNTINUHCSA-K
SMILES[Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ir 37.5% min
It is used as a photoreducing agent, a precursor for preparing Ir-C thin film and a catalyst for hydrogenation of sulfur containing compounds. Used in various catalysis reactions in organic synthesis, including carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only