Thermo Scientific Chemicals

Palladium(II) 2,4-pentanedionate, Pd 34.7%, Thermo Scientific Chemicals

Catalog number: 010517.06
5 g, Each
Thermo Scientific Chemicals

Palladium(II) 2,4-pentanedionate, Pd 34.7%, Thermo Scientific Chemicals

Catalog number: 010517.06
5 g, Each
Quantity
Catalog number: 010517.06
also known as 010517-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14024-61-4
IUPAC Name
palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C10H14O4Pd
InChI Key
JKDRQYIYVJVOPF-FDGPNNRMSA-L
SMILES
[Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Appearance (Color)
White to yellow to orange
Form
Powder
Assay (unspecified)
Palladium (Pd): 33.9-35.3%

Description

It is used in carbonylation, oligomerization, and as a catalyst precursor for various synthetic organic transformations, for example in the preparation of complexes to efficiently catalyze Heck reaction. Useful in the decarboxylative cross-coupling of arylcarboxylic acids with aryl halides, in the preparation of monodisperse nanoparticles of CuPd alloy and in several cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text