Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals
Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

For Use As Dehydrating and Chlorinating Agent, Catalyst
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
4 x 500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010523.36
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS10026-13-8
IUPAC Namepentachloro-λ⁵-phosphane
Molecular FormulaCl5P
InChI KeyUHZYTMXLRWXGPK-UHFFFAOYSA-N
SMILESClP(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (Titration ex Chloride)≥97.5% (U.K. specification)
Assay from Suppliers CofA≥97.5% (U.S. specification)
CommentPurchased in U.K. and U.S.
Impurity contentPCl₃ ≤0.5% (U.S. specification)
View more

Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only