Thermo Scientific Chemicals

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 010523.57
4 x 500 g, Each
Thermo Scientific Chemicals

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

Catalog number: 010523.57
4 x 500 g, Each
Quantity
Catalog number: 010523.57
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10026-13-8
IUPAC Name
pentachloro-λ⁵-phosphane
Molecular Formula
Cl5P
InChI Key
UHZYTMXLRWXGPK-UHFFFAOYSA-N
SMILES
ClP(Cl)(Cl)(Cl)Cl
Appearance (Color)
Yellow
Assay (Titration ex Chloride)
≥97.5% (U.K. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Comment
Purchased in U.K. and U.S.
Impurity content
PCl₃ ≤0.5% (U.S. specification)

Description

Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text