Thermo Scientific Chemicals

Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals

Catalog number: 010524.18
50 g, Each
Thermo Scientific Chemicals

Phosphorus(V) oxide, ACS, 98% min, Thermo Scientific Chemicals

Catalog number: 010524.18
50 g, Each
Quantity
Catalog number: 010524.18
also known as 010524-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-56-3
IUPAC Name
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular Formula
O5P2
InChI Key
DLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILES
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Assay (unspecified)
98.0% P₂O₅
Insoluble material
0.02%
Impurity content
NH₄ 0.01%
Impurity content
P₂O₃ P.T. (limit about 0.02%)
Trace Metals
(as Pb) 0.01%

Description

Used as a drying and dehydrating agent, condensation reagent in organic synthesis and a laboratory reagent. Used in sugar refining, and in fire extinguishing. Phosphorus pentoxide in DMSO forms Onodera reagent, which oxidizes alcohols. Capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text