Thermo Scientific Chemicals

Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals

Catalog number: 010525.A1
1 kg, Each
Thermo Scientific Chemicals

Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals

Catalog number: 010525.A1
1 kg, Each
Quantity
Catalog number: 010525.A1
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10025-87-3
IUPAC Name
phosphoroyl trichloride
Molecular Formula
Cl3OP
InChI Key
XHXFXVLFKHQFAL-UHFFFAOYSA-N
SMILES
ClP(Cl)(Cl)=O
Refractive Index
1.460-1.4620 @ 20°C (UK sourced material only)
Appearance (Color)
Clear
Color (APHA)
≤40
Assay from Supplier's CofA
≥98.5%
Form
Liquid

Description

Phosphorus(V) oxychloride is widely used as a dehydrating agent and a chlorinating agent. Useful for the conversion of amide to nitriles, Bischler-Napieralski reaction, Vilsmeier Haak reaction, and other reactions producing commercially important intermediates. Acts a source of liquid phosphorus in semiconductor industry. Manufacture of trialkyl phosphate esters and triarylphosphate esters, including commercially popular tricresyl phosphate,which is useful as a plasticizer, flame retardant, gasoline additive, hydrolytic fluid, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text