Thermo Scientific Chemicals

Platinum(II) 2,4-pentanedionate, Pt 48.0% min, Thermo Scientific Chemicals

Catalog number: 010526.03
1 g, Each
Thermo Scientific Chemicals

Platinum(II) 2,4-pentanedionate, Pt 48.0% min, Thermo Scientific Chemicals

Catalog number: 010526.03
1 g, Each
Quantity
Catalog number: 010526.03
also known as 010526-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15170-57-7
IUPAC Name
platinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C10H14O4Pt
InChI Key
KLFRPGNCEJNEKU-FDGPNNRMSA-L
SMILES
[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Assay (unspecified)
Pt 48.0% min

Description

Used for photoactivated hydrosilylation reactions between vinyl and silane groups; in the deposition of platinum metal and in the preparation of platinum nanotubes. Precursor for FePt superlattice magnetic nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text