Thermo Scientific Chemicals

Potassium pentachloronitrosylruthenate(II), Ru 25.8%, Thermo Scientific Chemicals

Catalog number: 010534.03
1 g, Each
Thermo Scientific Chemicals

Potassium pentachloronitrosylruthenate(II), Ru 25.8%, Thermo Scientific Chemicals

Catalog number: 010534.03
1 g, Each
Quantity
Catalog number: 010534.03
also known as 010534-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14854-54-7
IUPAC Name
dipotassium pentachloro(nitroso)rutheniumbis(ylium)
Molecular Formula
Cl5K2NORu
InChI Key
ZLWZLQOGXCFPCK-UHFFFAOYSA-I
SMILES
[K+].[K+].Cl[Ru++](Cl)(Cl)(Cl)(Cl)N=O
Assay (unspecified)
Ru 25.8% min

Description

Used as precipitation agent in Heterobimetallic coordination polymers involving 3d metal complexes and heavier transition metals cyanometallates. Ruthenium complexes are presently an object of great attention in the field of medicinal chemistry, as antitumor agents with selective antimetastatic properties and low systemic toxicity. Ruthenium compounds appear to penetrate reasonably well the tumor cells and bind effectively to DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text