Thermo Scientific Chemicals

Potassium tetranitroplatinate(II), Pt 42.6% min, Thermo Scientific Chemicals

Catalog number: 010538.03
1 g, Each
Thermo Scientific Chemicals

Potassium tetranitroplatinate(II), Pt 42.6% min, Thermo Scientific Chemicals

Catalog number: 010538.03
1 g, Each
Quantity
Catalog number: 010538.03
also known as 010538-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13815-39-9
IUPAC Name
platinum(2+) dipotassium tetranitrite
Molecular Formula
K2N4O8Pt
InChI Key
GQUAVGOGNMMUFS-UHFFFAOYSA-J
SMILES
[K+].[K+].[Pt++].[O-]N=O.[O-]N=O.[O-]N=O.[O-]N=O
Assay (unspecified)
Pt 42.6% min

Description

Potassium tetranitroplatinate(II) is used widely as laboratory chemicals and in manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text