Thermo Scientific Chemicals

Tetraammineplatinum(II) chloride monohydrate, Thermo Scientific Chemicals

Catalog number: 010544.03
1 g, Each
Thermo Scientific Chemicals

Tetraammineplatinum(II) chloride monohydrate, Thermo Scientific Chemicals

Catalog number: 010544.03
1 g, Each
Quantity
Catalog number: 010544.03
also known as 010544-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13933-33-0
IUPAC Name
platinum(2+) tetraamine hydrate dichloride
Molecular Formula
Cl2H14N4OPt
InChI Key
GWBDNMCYHWRSOH-UHFFFAOYSA-L
SMILES
N.N.N.N.O.[Cl-].[Cl-].[Pt++]
Assay (unspecified)
Pt 54-56% min

Description

Employed in the synthesis of cisplatin, and platinum included carbon - carbon materials with metal/metal oxide nanoparticles. Useful for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text