Thermo Scientific Chemicals

Tetracarbonyldi-^m-chlorodirhodium(I), Rh 50.1-52.9%, Thermo Scientific Chemicals

Catalog number: 010547.01
0.25 g, Each
Thermo Scientific Chemicals

Tetracarbonyldi-^m-chlorodirhodium(I), Rh 50.1-52.9%, Thermo Scientific Chemicals

Catalog number: 010547.01
0.25 g, Each
Quantity
Catalog number: 010547.01
also known as 010547-01
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14523-22-9
IUPAC Name
bis(λ¹-rhodium(1+)) tetrakis(methanidylidyneoxidanium) dichloride
Molecular Formula
C4Cl2O4Rh2
InChI Key
FGKDXBICTVUSPK-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Rh+].[Rh+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Assay (unspecified)
Rh 50.1-52.9%

Description

It is used widely as a chemical for research purposes as well as it acts as a a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text