Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals

Catalog number: 010548.04
2 g, Each
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals

Catalog number: 010548.04
2 g, Each
Quantity
Catalog number: 010548.04
also known as 010548-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14221-01-3
IUPAC Name
tetrakis(triphenylphosphane) palladium
Molecular Formula
C72H60P4Pd
InChI Key
NFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
Pd 9% min
Total Metal Impurities
0.2% max.
Appearance (Color)
Yellow to yellow/green
Form
Powder

Description

Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text