Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)platinum(0), Pt 15.2% min, Thermo Scientific Chemicals

Catalog number: 010549.06
5 g, Each
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)platinum(0), Pt 15.2% min, Thermo Scientific Chemicals

Catalog number: 010549.06
5 g, Each
Quantity
Catalog number: 010549.06
also known as 010549-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14221-02-4
IUPAC Name
tetrakis(triphenylphosphane) platinum
Molecular Formula
C72H60P4Pt
InChI Key
SYKXNRFLNZUGAJ-UHFFFAOYSA-N
SMILES
[Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Assay (unspecified)
Pt 15.2% min

Description

Tetrakis(triphenylphosphine)platinum(0), Pt is widely used in hydrosilation and oxidation. In Catalysts, Metal & Specialty Catalysts, Products & Industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text