Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%, Thermo Scientific Chemicals
Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.5 g
5 g
Catalog number 010553.03
also known as 010553-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS15004-86-1
IUPAC Namerhodium(3+) tris(ethane-1,2-diamine) trihydrate trichloride
Molecular FormulaC6H30Cl3N6O3Rh
InChI KeyHHSLUQMEKOPKJG-UHFFFAOYSA-K
SMILESO.O.O.[Cl-].[Cl-].[Cl-].[Rh+3].NCCN.NCCN.NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or fused lumps
Assay (unspecified)Rh: ≥23.15%
Used as precursor for supported RH catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only