Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals
Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 010567.18
also known as 010567-18
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
Add to cart
Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number010567.18
Price (USD)/ Each
161.65
Online exclusive
179.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7488-54-2
IUPAC Namesulfuric acid rubidium
Molecular FormulaH2O4RbS
InChI KeyXNNUAZGCOYTNQE-UHFFFAOYSA-N
SMILES[Rb].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Rubidium sulfate is used in hematology stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only