Thermo Scientific Chemicals

Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals

Catalog number: 010568.03
1 g, Each
Thermo Scientific Chemicals

Ruthenium(III) 2,4-pentanedionate, Ru 24% min, Thermo Scientific Chemicals

Catalog number: 010568.03
1 g, Each
Quantity
Catalog number: 010568.03
also known as 010568-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14284-93-6
IUPAC Name
ruthenium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21O6Ru
InChI Key
RTZYCRSRNSTRGC-LNTINUHCSA-K
SMILES
[Ru+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Concentration
Ru 24% min.
Assay (unspecified)
Ru 24% min

Description

Precursor for the preparation of other compounds of ruthenium. Ruthenium(III) 2,4-pentanedionate is employed as a recyclable catalyst for certain organic transformations like the acetylation of phenols, alcohols, and amines under neat conditions. It is also used as a homogeneous catalyst, for example, in the hydrolysis of sodium borohydride and regiospecific tritiation of aromatic carboxylic acids. It is also used as a catalyst for enantioselective hydrogenation of acids such as aryl acrylic acid and aryl propenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text