Nickel powder, -60+170 mesh, 99.7% (metals basis), Thermo Scientific Chemicals
Nickel powder, -60+170 mesh, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel powder, -60+170 mesh, 99.7% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010579.36
also known as 010579-36
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
Add to cart
Nickel powder, -60+170 mesh, 99.7% (metals basis), Thermo Scientific Chemicals
Catalog number010579.36
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Particle Size-60+170 mesh
Appearance (Color)Grey
Total Metal Impurities≤0.3%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only