Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium foil, 0.127mm (0.005in) thick, annealed, 99.5% (metals basis), Thermo Scientific Chemicals

Form: Approx.10.3g/100 x 125mm, Temper: soft
Have Questions?
Change viewbuttonViewtableView
Quantity:
125 x 2000 mm
100 x 125 mm
125 x 500 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010594.IB
Price (USD)/ Each
-
Quantity:
125 x 2000 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-67-7
IUPAC Namezirconium
Molecular FormulaZr
InChI KeyQCWXUUIWCKQGHC-UHFFFAOYSA-N
SMILES[Zr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only