Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 010601.06
also known as 010601-06
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
Add to cart
Selenium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Catalog number010601.06
Price (USD)/ Each
41.65
Online exclusive
45.80 
Save 4.15 (9%)
-
Add to cart
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark grey to black
FormPowder
Total Metal Impurities≤0.5%
Particle Size-325 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only