Selenium shot, amorphous, 2-6mm (0.08-0.2in), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Selenium shot, amorphous, 2-6mm (0.08-0.2in), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Selenium shot, amorphous, 2-6mm (0.08-0.2in), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 010603.30
also known as 010603-30
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
Add to cart
Selenium shot, amorphous, 2-6mm (0.08-0.2in), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010603.30
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7782-49-2
IUPAC Nameselenium
Molecular FormulaSe
InChI KeyBUGBHKTXTAQXES-UHFFFAOYSA-N
SMILES[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only