Tellurium powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Tellurium powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tellurium powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010605.18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS13494-80-9
IUPAC Nametellurium
Molecular FormulaTe
InChI KeyPORWMNRCUJJQNO-UHFFFAOYSA-N
SMILES[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -200 mesh
Total Metal Impurities≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only