Thermo Scientific Chemicals

Tellurium(IV) oxide, tech., Thermo Scientific Chemicals

Catalog number: 010608.30
250 g, Each
Thermo Scientific Chemicals

Tellurium(IV) oxide, tech., Thermo Scientific Chemicals

Catalog number: 010608.30
250 g, Each
Quantity
Catalog number: 010608.30
also known as 010608-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-07-3
IUPAC Name
(oxo-λ⁴-tellanylidene)oxidane
Molecular Formula
O2Te
InChI Key
LAJZODKXOMJMPK-UHFFFAOYSA-N
SMILES
O=[Te]=O
Impurity content
1% max.

Description

It is used as an acousto-optic material and a conditional glass former. The glasses of tellurium oxide have high refractive indices. Owing to the transmission into mid-infrared part of the electromagnetic spectrum, tellurium oxide is of technological interest for optical waveguides. It is also employed in optical fibre amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text