Thermo Scientific Chemicals

Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010622.18
50 g, Each
Thermo Scientific Chemicals

Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010622.18
50 g, Each
Quantity
Catalog number: 010622.18
also known as 010622-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-13-6
IUPAC Name
aluminium(3+) hexahydrate trichloride
Molecular Formula
AlCl3H12O6
InChI Key
JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
Appearance (Color)
White
Form
Crystals or powder
Total Metal Impurities
≤0.0005% (5 ppm)
Assay (special)
?2.5 % of Theoretical value (Al)

Description

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text