Thermo Scientific Chemicals

Ammonium chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010632.A1
1 kg, Each
Thermo Scientific Chemicals

Ammonium chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010632.A1
1 kg, Each
Quantity
Catalog number: 010632.A1
also known as 010632-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-02-9
IUPAC Name
amine hydrochloride
Molecular Formula
ClH4N
InChI Key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES
N.Cl
Total Metal Impurities
0.001% max.

Description

Ammonium chloride finds its use as a flux in preparing metals for galvanizing, soldering or tin coating. While mainly referred as a nitrogen source fertilizer, it is also used in cleaning tip of soldering iron, hair shampoo as a thickening agent, some batteries as electrolyte, tanning, textile printing, dyeing, and cleaning products. In laboratories it is used to produce cooling baths and for producing buffers. Along with certain metals, it is used for effecting certain reductions in organic synthesis. It can be used as a precursor to ammonium cyanate, which is employed in the Wöhler synthesis of urea. It facilitates several organic transformations in a mild and efficient manner - for example in Ulmann coupling, cross-aldol condensation, Claisen rearrangement, reduction, thia-Michael addition and synthesis of several monocyclic, fused or spiro bicyclic nitrogen heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text