Ammonium dihydrogen phosphate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010633.14
also known as 010633-14
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
Add to cart
Ammonium dihydrogen phosphate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010633.14
Price (USD)/ Each
127.65
Online exclusive
142.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Crystals of this compound are used in transducers as an alternative to Quartz and Rochelle salt. Monoammonium phosphate is one of the components of agricultural fertilizers. It supplies elements nitrogen and phosphorus to the soil. It is also a component of some dry chemical fire extinguishers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only