Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
50 g
100 g
Catalog number 010641.06
also known as 010641-06
Price (USD)/ Each
41.65
Online exclusive
46.20 
Save 4.55 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
46.20 
Save 4.55 (10%)
Add to cart
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010641.06
Price (USD)/ Each
41.65
Online exclusive
46.20 
Save 4.55 (10%)
-
Add to cart
Chemical Identifiers
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)?2.5 % of Theoretical value (Sb)
Appearance (Color)White
FormPowder
Total Metal Impurities≤0.001% (10 ppm)
Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only