Thermo Scientific Chemicals

Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010641.22
100 g, Each
Thermo Scientific Chemicals

Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010641.22
100 g, Each
Quantity
Catalog number: 010641.22
also known as 010641-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-64-4
IUPAC Name
diantimony(3+) trioxidandiide
Molecular Formula
O3Sb2
InChI Key
GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sb+3].[Sb+3]
Assay (special)
?2.5 % of Theoretical value (Sb)
Appearance (Color)
White
Form
Powder
Total Metal Impurities
≤0.001% (10 ppm)

Description

Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text