Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 010645.36
also known as 010645-36
Price (USD)/ Each
1,547.65
Online exclusive
1719.00 
Save 171.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
1,547.65
Online exclusive
1719.00 
Save 171.35 (10%)
Add to cart
Barium carbonate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number010645.36
Price (USD)/ Each
1,547.65
Online exclusive
1719.00 
Save 171.35 (10%)
-
Add to cart
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Barium carbonate is frequently employed in ceramic industry as a component of glazes. It is added to clay to precipitate some soluble salts while manufacturing bricks, tiles and potteries. Barium carbonate is mainly used as a raw material in the electrical engineering industries for glass production, and as a raw material in the magnet manufacturing industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only